Ethylene glycol, 99.8%, anhydrous, AcroSeal™
Ethylene glycol, 99.8%, anhydrous, AcroSeal™
Ethylene glycol, 99.8%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

Ethylene glycol, 99.8%, anhydrous, AcroSeal™

CAS: 107-21-1 | C2H6O2 | 62.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 433811000
Price (EUR)
59,65
Online exclusive
66,20
Save 6,55 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
59,65
Online exclusive
66,20
Save 6,55 (10%)
Each
Chemical Identifiers
CAS107-21-1
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=99.75 %
Water=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only