n-Pentane, 95%, technical grade
n-Pentane, 95%, technical grade
n-Pentane, 95%, technical grade
Thermo Scientific Chemicals

n-Pentane, 95%, technical grade

CAS: 109-66-0 | C5H12 | 72.15 g/mol
数量:
5 L
25 L
パッケージ:
Metal Drum
製品番号(カタログ番号) 434030250
価格(JPY)
-
数量:
25 L
パッケージ:
Metal Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3570 to 1.3590 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only