Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous
Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous
Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous
Thermo Scientific Chemicals

Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous

CAS: 54761-04-5 | C3F9O9S3Yb | 620.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 434050050
Price (USD)
108.65
Online exclusive
121.00
Save 12.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
108.65
Online exclusive
121.00
Save 12.35
Each
Chemical Identifiers
CAS54761-04-5
IUPAC Nameytterbium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Yb
InChI KeyAHZJKOKFZJYCLG-UHFFFAOYSA-K
SMILES[Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Complexometric26.8 to 29.0 % (Yb)
Infrared spectrumConforms
Total conc. of imp.as REO=<1000 ppm
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only