Product Image
Applied Biosystems™

Acetic Anhydride/Pyridine/THF, Capping Formula A

Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
43408604 x 450 mL
43408654 x 180 mL
43473814 x 2 L
Número de catálogo 4340860
Precio (CLP)
-
Cantidad:
4 x 450 mL
This acetic anhydride⁄tetrahydrofolate⁄pyridine (Cap A) solution comes in Trap-Pak™ bags. Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to efficiently adsorb water. Use Trap-Pak™ bags to reduce and maintain a low water content in acetonitrile and tetrazole (activator) solutions.

• Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
• Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.

Tested
Our high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Nombre del producto químico o materialAcetic anhydride/pyridine/tetrahydrofuran
Para utilizar con (aplicación)Síntesis de oligos
Para utilizar con (equipo)Sintetizador de ADN 3900
FormularioLíquido
Tipo de productoReactivos de síntesis de oligonucleótidos
Cantidad4 x 450 mL
Tipo de reactivoDisolvente o reactivo general
Tipo de muestraDNA
Unit SizeEach
Contenido y almacenamiento
4 x 450 mL