Product Image
Applied Biosystems™

Iodine-Water-Pyridine-Tetrahydrofuran Oxidizer Solution (0.02 M)

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43408624 x 450 mL
401732200 mL
401632450 mL
43408674 x 180 mL
43473804 x 2 L
Catalog number 4340862
Price (USD)
714.00
4 x 450 mL
Quantity:
4 x 450 mL
Price (USD)
714.00
4 x 450 mL
This Iodine⁄Water⁄Pyridine⁄Tetrahydrofuran (oxidizer) solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.

• Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
• Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
• Reagent kits are available for your convenience, and save you time and money.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.

For Research Use Only. Not for use in diagnostics procedures.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPyridine Tetrahydrofuran
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormLiquid
Product TypeOligonucleotide Synthesis Reagents
Quantity4 x 450 mL
Reagent TypeSolvent or General Reagent
Sample TypeDNA
Unit Size4 x 450 mL
Contents & Storage
4 × 450 ml