N-Methyl-1,2-phenylenediamine, 97%
N-Methyl-1,2-phenylenediamine, 97%
N-Methyl-1,2-phenylenediamine, 97%
Thermo Scientific Chemicals

N-Methyl-1,2-phenylenediamine, 97%

CAS: 4760-34-3 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 434230250
Price (EUR)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4760-34-3
IUPAC NameN1-methylbenzene-1,2-diamine
Molecular FormulaC7H10N2
InChI KeyRPKCLSMBVQLWIN-UHFFFAOYSA-N
SMILESCNC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear yellow to red to dark brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only