Are there any His-tags on Streptavidin (Cat. No. 434302)?
This Streptavidin is a native protein from Streptomyces avidinii, therefore there are no His-tags on it.
Search
View additional product information for Streptavidin, lyophilized powder - FAQs (434302, 434301)
3 product FAQs found