Product Image
Invitrogen™

Streptavidin - Cy5

Streptavidin - Cy5
Have Questions?
Catalog NumberQuantity
4343161 mL
Catalog number 434316
Price (CLP)
314.806
Each
Quantity:
1 mL
Price (CLP)
314.806
Each
Reactivity: CY5-SA reacts with biotin.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mg/mL
Product TypeStreptavidin Conjugate (fluorescent)
Quantity1 mL
Shipping ConditionWet Ice
ConjugateCy5
FormLiquid
Product LineCy
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

Is there a crosslinker used in Streptavidin-FITC (Cat. No. 434311) and Streptavidin-Cy5 (Cat. No. 434316) conjugates?

There is no crosslinker as the dyes themselves are made to react directly with the streptavidin protein.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many molecules of protein and dye are in 1 mg of Streptavidin-Cy5 (Cat. No. 434316)?

There are about 3.993 x 10^16 molecules of dye and 1.141 x 10^16 of streptavidin in 1 mg of Streptavidin-Cy5.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the dye-protein ratio for Streptavidin-Cy5 (Cat. No. 434316)?

The target dye to protein ratio for Streptavidin-Cy5 (Cat. No. 434316) is 4:1. The average number of dye molecules on each streptavidin molecule is between 3 and 4.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.