5-Amino-2-chloropyrimidine, 95%
5-Amino-2-chloropyrimidine, 95%
5-Amino-2-chloropyrimidine, 95%
Thermo Scientific Chemicals

5-Amino-2-chloropyrimidine, 95%

CAS: 56621-90-0 | C4H4ClN3 | 129.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 434320050
Price (EUR)
423,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
423,00
Each
Chemical Identifiers
CAS56621-90-0
IUPAC Name2-chloropyrimidin-5-amine
Molecular FormulaC4H4ClN3
InChI KeyDZBKIOJXVOECRA-UHFFFAOYSA-N
SMILESNC1=CN=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
Appearance (Color)Yellow to brown
GC>=94.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only