Streptavidin - Agarose
Invitrogen™

Streptavidin - Agarose

Estreptavidina (agarosa)
Have Questions?
Número de catálogoCantidad
4343412 mL
Número de catálogo 434341
Precio (MXN)
-
Cantidad:
2 mL
Pedido a granel o personalizado

Reactivity: SA-Seph 4B reacts with biotin.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Concentración1 mg de estreptavidina/ml de gel de Sepharose
Tipo de productoConjugado de estreptavidina (Fluorescente)
Cantidad2 mL
Condiciones de envíoHielo húmedo
FormularioSuspensión líquida
Unit SizeEach
Contenido y almacenamiento
Almacenar en el refrigerador (2–8° C).

Preguntas frecuentes

How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Entamoeba histolytica Cell Surface Calreticulin Binds Human C1q and Functions in Amebic Phagocytosis of Host Cells.
Authors:Vaithilingam A, Teixeira JE, Miller PJ, Heron BT, Huston CD
Journal:Infect Immun
PubMed ID:22473608
'Phagocytosis of host cells is characteristic of tissue invasion by the intestinal ameba Entamoeba histolytica, which causes amebic dysentery and liver abscesses. Entamoeba histolytica induces host cell apoptosis and uses ligands, including C1q, on apoptotic cells to engulf them. Two mass spectrometry analyses identified calreticulin in amebic phagosome preparations, and, ... More