Ammonium Sulfate, 99%, extra pure
Ammonium Sulfate, 99%, extra pure
Ammonium Sulfate, 99%, extra pure
Thermo Scientific Chemicals

Ammonium Sulfate, 99%, extra pure

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 434380010
Price (USD)
53.65
Online exclusive
59.20
Save 5.55
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
53.65
Online exclusive
59.20
Save 5.55
Each
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<0.002 %
Appearance (Form)Crystals
Iron (Fe)=<0.002 %
pH5.0 to 6.0
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only