Avidin, HRP conjugate
Invitrogen™

Avidin, HRP conjugate

Avidin, HRP conjugate
Have Questions?
Catalog NumberQuantity
4344235 mg
Catalog number 434423
Price (MXN)
-
Quantity:
5 mg

Reactivity: HRP-Avidin reacts with biotin.

Applications: Immunohistochemistry, ELISA

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Label or DyeHRP (Horseradish Peroxidase)
Quantity5 mg
Shipping ConditionWet Ice
ConjugateHRP
FormLiquid
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

Can you provide the shelf life for Avidin, HRP conjugate (Cat. No. 434423)?

The Avidin, HRP conjugate (Cat. No. 434423) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Subtyping lymphocytes in peripheral blood by immunoperoxidase labeling and light scatter/absorption flow cytometry.
Authors:Kim YR, Martin G, Paseltiner L, Ansley H, Ornstein L, Kanter RJ
Journal:Clin Chem
PubMed ID:3896568
'Lymphocyte subpopulations in a whole-blood sample can be detected by adapting mouse monoclonal antibodies (MAbs) and peroxidase (EC 1.11.1.7) labeling to a flow cytometer equipped with a tungsten-halogen light source and scatter/absorption optics (Technicon H6000). In the optimized cytochemical conditions each cell population generates a distinct, well-separated cluster, for accurate ... More