4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%
4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%
4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%
Thermo Scientific Chemicals

4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%

CAS: 55-55-0 | C14H18N2O2·H2SO4 | 344.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 434441000
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.90
Save 8.25
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.90
Save 8.25
Each
Chemical Identifiers
CAS55-55-0
IUPAC Namebis(4-(methylamino)phenol); sulfuric acid
Molecular FormulaC14H20N2O6S
InChI KeyZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Suitability(for determ. of phosphate) Passes test
Appearance (Color)White to off-white
AppearanceDiscolors in air
Appearance (Form)Crystalline powder
Titration99.0 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only