Large Volume Upgrade Starter Kit

Catalog number: 4344440

Ambion™  Related applications: RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Large Volume Upgrade Starter Kit upgrades the ABI PRISM® 6100 Nucleic Acid PrepStation for sample preparation of large volume Tempus™ Blood RNA Tubes.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 6100 Nucleic Acid PrepStation
Product Line: Applied Biosystems™
Product Size: 1 kit
Shipping Condition: Room Temperature
Type: Adapter, Containers & Vessels, Filter, Plate, Tubes & Vials

Contents & storage

The kit contains a volume adapter plate, elution vial, cap plugs, reusable tube collection plates, Microcentrifuge tubes, RNA filters, and two different sizes of reservoirs.

Documents

Manuals & protocols