Try PowerTrack SYBR Green Master Mix - our newest formulation simplifies your workflow by minimizing primer optimization and reducing pipetting errors with an optional sample tracking dye.
SYBR™ Green Universal Master Mix

SYBR™ Green Universal Master Mix

Notification of updated SYBR Green master mix formulation: PowerTrack SYBR Green Master Mix is our newest formulation designed to greatlyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43444631 x 1 mL
43091551 x 5 mL
43643442 x 5 mL
43643465 x 5 mL
431270410 x 5 mL
43349731 x 50 mL
Catalog number 4344463
Price (USD)
129.93
Special offer
Online exclusive
Ends: 11-Dec-2026
183.00
Save 53.07
Each
Quantity:
1 x 1 mL
Price (USD)
129.93
Special offer
Online exclusive
Ends: 11-Dec-2026
183.00
Save 53.07
Each

Notification of updated SYBR Green master mix formulation: PowerTrack SYBR Green Master Mix is our newest formulation designed to greatly simplify your work and deliver high sensitivity and specificity. It comes with an optional yellow dye that can be added to your DNA sample for pipetting QC. Yellow sample added to blue master mix yields green qPCR reaction. Visually confirm that your DNA sample was added to master mix. PowerTrack SYBR Green Master Mix is compatible with a wide range of primer melting temperatures (55°C to 65°C) and concentrations to minimize primer optimization work.

----------------------------------------------

SYBR Green PCR Master Mix
Everything you need for SYBR Green dye–based PCR amplification and detection in a convenient, single-tube format. Applied Biosystems SYBR Green PCR Master Mix combines SYBR Green I dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.

  • Premixed components stored at 2–8°C significantly reduce assay setup time
  • SYBR Green I dye detects double-stranded DNA, so specific probes are not required
  • AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
  • dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
  • Proprietary buffer enhancements ensure performance and reliability

Maximum flexibility and convenience
Applied Biosystems SYBR Green PCR Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan probes are required. SYBR Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR. The hot-start enzyme AmpliTaq Gold DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity. Passive Internal Reference 1 is provided to normalize non-PCR–related fluorescence fluctuations. This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation. SYBR Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, AB 7900 HT 384-Well System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System, GeneAmp™ 5700
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions40 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineSYBR
Product TypeReal Time PCR SYBR Master Mix
Quantity1 x 1 mL
Sample TypeDNA
Shipping ConditionWet Ice
Sufficient For40 Reactions of 50 μL
Target SpecificityNot Target-Specific
TechniqueFluorescence Intensity
Volume1 x 1 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
FormConcentrated
GC-Rich PCR PerformanceHigh
Label or DyeSYBR Green
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
SYBR™ Green PCR Master Mix in a 1 mL vial. Sufficient reagents provided for 40 reactions based on a 50 μl reaction volume. Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What is the difference in sensitivity between TaqMan chemistry vs. SYBR Green reagent chemistry?

Sensitivity can actually be equivalent when using TaqMan chemistry and SYBR Green reagent chemistry. It might seem that a TaqMan assay with fluorescent signal generated by a sequence-specific probe would always be more sensitive than a SYBR Green reagent assay, but a poorly designed TaqMan assay could theoretically be less specific than a well-designed SYBR Green reagent assay. However, the potential for detection of primer dimers and non-specific products using SYBR Green chemistry is more likely to result in loss of sensitivity when attempting to quantitate lower copy numbers.

For more information on Real-Time PCR chemistries, please refer to the following Application Notes, which you can find on our website through Technical Resources, or by entering the titles in the main Search field: “Real-Time PCR Vs. Traditional PCR”, “Essentials of Real Time PCR”, and “Selection of Reagents for Real-Time PCR”.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

What are the key differences between a TaqMan MGB probe and a TaqMan TAMRA dye-labeled probe?

The TaqMan MGB probes contain the following features:
1) A fluorescent reporter at the 5' end
2) A nonfluorescent quencher at the 3' end. Because the quencher does not fluoresce, the real-time instruments can measure the reporter dye contributions more precisely.
3) A minor groove binder at the 3' end. The minor groove binder increases the melting temperature (Tm) of probes, allowing the use of shorter probes.

In general, the TaqMan MGB probes exhibit great differences in Tm values between matched and mismatched probes, which provides more accurate allelic discrimination and makes for a more sensitive real-time assay. Mismatches between a probe and allele, or target, reduce the efficiency of probe hybridization in a measurable way, which is especially important in SNP Genotyping assays. Furthermore, AmpliTaq Gold DNA polymerase is more likely to displace the mismatched probe rather than cleave it to release reporter dye. More information about TaqMan MGB probes can be found in the User Bulletin entitled "Primer Express Version 1.5 and TaqMan MGB Probes for Allelic Discrimination." You can find a copy on our website by entering this title in the main search field.

When using SYBR Green chemistry on an Applied Biosystems Real-Time PCR instrument, how do I change settings to reflect that there is no TaqMan probe being used in the reaction?

Refer to the product manual for your instrument and software for specifics, but in general you will want to change the Quencher value to None.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

How can RNA standards be generated to perform absolute quantitation for RNA targets?

It is generally not possible to use DNA as a standard for absolute quantitation of RNA because there is no control for the efficiency of the reverse transcription step. Therefore, in-vitro transcribed RNA is commonly used to prepare standards for the absolute quantitation of RNA targets. This would involve the cloning of the target of interest into an in-vitro transcription plasmid, performing in-vitro transcription, then purifying the resulting cRNA so that the DNA plasmid cannot serve as a PCR template. Concentration is measured by A260 and converted to the number of copies using the molecular weight of the RNA.

Relative quantitation of gene expression methods require less up-front preparation and provide a fold-change value instead of an absolute quantity result. For many researchers, absolute quantities are not a necessary parameter to measure, and therefore relative quantitation is a much more attractive approach to studying gene expression via real-time PCR. For more information on relative quantitation of gene expression, please refer to our Technical Reference Library in the Technical Resources section of our website.