Tris(hydroxymethyl)aminomethane acetate salt, 99%
Tris(hydroxymethyl)aminomethane acetate salt, 99%
Tris(hydroxymethyl)aminomethane acetate salt, 99%
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane acetate salt, 99%

CAS: 6850-28-8 | C4H11NO3·C2H4O2 | 181.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 434500250
Price (USD)
71.40
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
71.40
Each
Chemical Identifiers
CAS6850-28-8
IUPAC Name1,3-dihydroxy-2-(hydroxymethyl)propan-2-aminium acetate
Molecular FormulaC6H15NO5
InChI KeyPIEPQKCYPFFYMG-UHFFFAOYSA-N
SMILESCC([O-])=O.[NH3+]C(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 290 nm A: =<0.5
Appearance (Color)White
Appearance (Form)Crystalline powder
Water=<1.0 %
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only