beta-Alanine-tert-butylester hydrochloride, 98%
beta-Alanine-tert-butylester hydrochloride, 98%
beta-Alanine-tert-butylester hydrochloride, 98%
Thermo Scientific Chemicals

beta-Alanine-tert-butylester hydrochloride, 98%

CAS: 58620-93-2 | C7H16ClNO2 | 181.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 434650010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS58620-93-2
IUPAC Nametert-butyl 3-aminopropanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyDOMTZTVJNZKUNX-UHFFFAOYSA-N
SMILESCl.CC(C)(C)OC(=O)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Elemental Analysis45.1 to 47.4 % C
Appearance (Form)Crystalline powder or solid
Elemental Analysis7.5 to 7.9 % N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only