Urea, 99+%, for biochemistry
Urea, 99+%, for biochemistry
Urea, 99+%, for biochemistry
Thermo Scientific Chemicals

Urea, 99+%, for biochemistry

CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 434720010
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.60
Save 13.95
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.60
Save 13.95
Each
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV(6 M in H2O)
UVat 280 nm A: =<0.06
Ammonium (NH4)=<0.05 %
Chloride (Cl)=<0.05 %
Loss on drying=<0.1 % (1 g, 105°C, 1 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only