This tetrazole⁄acetonitrile solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.
• Reagent concentrations are within our strict specifications, guaranteeing consistent quality. • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product. • Reagent kits are available for your convenience, and save you time and money.
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Nombre del producto químico o materialAcetonitrilo tetrazol
Para utilizar con (aplicación)Síntesis de oligos
Para utilizar con (equipo)Sintetizador de ADN 3900
FormularioLíquido
Tipo de productoReactivos de síntesis de oligonucleótidos
Cantidad4 x 2 L
Tipo de reactivoDisolvente o reactivo general
Tipo de muestraDNA
Unit Size4 x 2 L
Contenido y almacenamiento
Topfill consisting of four each of Tetrazole⁄Acetonitrile Part Number 401271 Store in a cool, dry, and well-ventilated location, appropriate for flammable lliquids.