Product Image
Applied Biosystems™

MicroSEQ™ D2 rDNA Fungal Sequencing Kit

This kit is the sequencing component of the MicroSEQ Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi.
Have Questions?
產品號碼Quantity
43474811 Kit
產品號碼 4347481
價格 (TWD)
-
Quantity:
1 Kit

This kit is the sequencing component of the MicroSEQ™ Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi. It includes the primers needed to sequence the PCR products generated using the PCR component. A protocol and Quick Reference Card are not included.

  • Sequence-based measurement ensures you get reproducible results in less than 24 hours.
  • Growth requirement-independent measurement enables you to identify fungi using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training; same work flow as the MicroSEQ™ Full Gene 16S rDNA Bacterial Identification Kit.

Specific and Easy to Use
Based on phylogenetic analysis of the D2 region of large-subunit ribosomal RNA gene sequences, this easy-to-use kit enables you to identify even the most difficult fungi, yeast, and oomycete plant pathogens. To ensure accurate and specific results, the kit's software compares your samples to a database of D2 sequences from over 1000 validated type species.

The MicroSEQ™ ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

For Research Use Only. Not for use in diagnostic procedures.

規格
Detection MethodCapillary Electrophoresis
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 9800 Fast Thermal Cycler
LibrariesFungal Library (> 1100 species), Customizable
Product LineMicroSEQ
Product TypeSequencing Kit
Quantity1 Kit
Sample TypePCR Products
Shipping ConditionDry Ice
SoftwareMicroSEQ™ ID Analysis Software
TargetD2 region of rRNA gene
Target Organism ClassFungi
TechniqueSBT (Sequence-Based Typing)
Workflow StepCycle Sequencing
FormatKit
Unit SizeEach
內容物與存放
The kit contains reagents sufficient for 55 sequencing reactions:
• D2 Forward Sequencing Mix
• D2 Reverse Sequencing Mix

Store at -15°C to -25°C

常見問答集 (常見問題)

Do you have stability information for the MicroSEQ D2 rDNA Fungal Sequencing Kit (Cat. No. 4347481) when subjected to temperature fluctuations?

The MicroSEQ D2 rDNA Fungal Sequencing Kit is stable if stored at -20 degrees C for 15-16 months. We do not have any information available for temperatures outside of this range.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.