MicroSEQ® Full Gene 16S rDNA PCR Kit, with protocol & quick reference card

Catalog number: 4349161

Applied Biosystems™  Related applications: Microbial Identification & Detection | Microbial Protein Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The MicroSEQ® Full Gene 16S rDNA PCR Kit is the PCR component of the MicroSEQ® 16S rDNA Full Gene Identification Kit, which provides an easy-to-use DNA sequence-based method to identify most bacteria. Use the PCR kit to amplify the bacterial 16S rRNA gene, for subsequent sequencing using the MicroSEQ® Full Gene 16S rDNA Sequencing Kit. This PCR kit does include a protocol and a Quick Reference Card.

• Sequence-based measurement ensures you get reproducible results in under 24 hours.
• Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
• Universal domain-level primers eliminate up-front optimization.
• MicroSEQ® ID Analysis Software ensures accurate species identification.
• Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
• Requires minimal training-similar work flow as the MicroSEQ® D2 LSU fungal kit and the MicroSEQ® 500 16S rDNA kit.

The MicroSEQ® ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems® MicroSEQ; System combines the advantages of MicroSEQ® ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ® bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Target Organism Class: Bacteria
Target Gene: 16S rRNA (full gene)
Validated Application: Polymerase Chain Reaction
Detection Method: Gel Electrophoresis
For Use With (Equipment): GeneAmp™ 9700, Veriti® Thermal Cycler
Sample Type (Specific): DNA (Genomic)
Technique: SBT (Sequence-Based Typing)
Library Type: Bacterial Library (> 2000 species), Customizable
Analysis Software: MicroSEQ® ID Analysis Software
Product Line: Applied Biosystems™, MicroSEQ®
Product Size: 1 kit
Target Molecule: DNA
Workflow Step: PCR Amplification

Contents & storage

Each kit contains:
3 tubes 2X PCR Master Mix (sufficient for 20 amplifications).
1 tube positive control DNA.
1 tube negative control DNA (water).
After first use, store at 2 to 6°C in a PCR clean room.

Documents

Manuals & protocols