Product Image
Applied Biosystems™

Sequence Detection System v2.4 Enterprise Edition

Sequence Detection System (SDS) Software v2.4.1 is the latest update in high-throughput gene expression and genotyping analysis software for useRead more
Have Questions?
Catalog NumberQuantity
43504901 software
Catalog number 4350490
Price (USD)
36,960.00
Each
Quantity:
1 software
Price (USD)
36,960.00
Each
Sequence Detection System (SDS) Software v2.4.1 is the latest update in high-throughput gene expression and genotyping analysis software for use with the Applied Biosystems™ 7900HT Fast Real Time PCR System.

Features of SDS Software:

• Assisted Tm Calling
• Ability to create and save analysis templates on a per detector basis for use with other plates
• Auto-analysis of plates containing analysis settings
• Improvements in amplification plot displays, including the ability to view plots by sample and target, as well as a toggle for linear/log display
• Ability to access the plate utility application from the main menu
• Ability to export the results or data of up to 50 plates at once, as well as a choice of results columns to export, using the batch export utility
• Ability to set up standard curve dilutions for AQ experiments with a wizard
• Ability to switch run documents from AQ to RQ format, and vice-versa

Standard and Enterprise Editions
The standard version of SDS Software for the 7900HT Fast Real-Time PCR System uses two automation software tools—Plate Utility and Automation Controller—to fully automate high-throughput applications for gene expression and SNP genotyping, requiring minimal user intervention.

The Enterprise Edition of SDS Software is a high-throughput application for storage, organization, and analysis of large amounts of gene expression and genotyping data on the 7900HT Fast Real-Time PCR System
• Upgrade to Oracle Database 11g
• Process-based quality flags for increased quality control, problem notification, filtering, and troubleshooting
• Improved review and data group tools
• Wizard-based plate document creation
• Support for plus/minus assays
• Re-engineered SDS Automation Controller
• Ability to save plates created using the template batch utility directly to the database

The Enterprise database allows users to process up to 5,000 x 384-well plates (384 markers total), which equates to 1.92 million genotypes. Using Relative Quantitation (RQ) Manager, customers can also process 200 x 384-well plates (96 detectors with quadruplicate data points per multiplexed endogenous control) which equates to 153,600 data points.

Supporting 21 CFR Part 11 Requirements
SDS Software Enterprise Edition is the only edition of SDS Software that assists in supporting 21 CFR part 11 requirements, providing security, auditing functions, electronic signatures, and electronic record-keeping. These features eliminate traditional paper records and reduce vulnerability to signature fraud and report misfiling, which increases the speed of information exchange and data searching. The results are reduced storage costs, better data integration, and superior product quality.

For Research Use Only. Not intended for any animal, human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Applied Biosystems 7900HT Fast Real Time PCR System
Format384-Well Plate
Product TypeSequence Detection System (SDS) Software
Quantity1 software
Software CategorySoftware
Software TypeReal-Time PCR Data Analysis
Unit SizeEach
Contents & Storage
1 Software Kit

Frequently asked questions (FAQs)

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.