Product Image
Applied Biosystems™

7900HT System Spectral Calibration Kit, Fast 96-well

This easy-to-use kit enables you to establish the pure dye spectra and multicomponent values for FAM™, JOE™, NED™, ROX™, SYBR™Read more
Have Questions?
Catalog NumberQuantity
43516531 kit
Catalog number 4351653
Price (USD)
982.00
Each
Quantity:
1 kit
Price (USD)
982.00
Each
This easy-to-use kit enables you to establish the pure dye spectra and multicomponent values for FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™ dyes on the 7900HT Fast Real-Time PCR System. The Applied Biosystems™ 7900 HT System Fast 96-Well Spectral Calibration Kit contains three Optical 96-Well Fast Thermocycling Plates: one preloaded and sealed background plate and two preloaded and sealed with eight separate dye standards.

• 96-well optical reaction plates preloaded with FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™ dyes allow quick and easy calibration of the Applied Biosystems™ 7900HT Fast Real-Time PCR System
• Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously
• Includes the Passive Reference I dye standard required for signal normalization in all real-time and endpoint reactions

Spend Your Time on Research, Not Instrument Calibration
This kit is easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7900HT Fast Real-Time PCR System. Insert the reaction plates during system installation, and the system collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeFAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green
Calibration TypeSpectral Calibration
Detection MethodFluorescence
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7900HT Fast System
Quantity1 kit
Shipping ConditionDry Ice
Product TypeSpectral Calibration Kit
Unit SizeEach
Contents & Storage
Contains one Background Plate and two spectral calibration plates (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™,and VIC™ dye standards).

Store at -15 to 25°C. (This product is light sensitive.)

Expiration date: 12 months from the date of delivery

Frequently asked questions (FAQs)

How can I convert a file from AQ to RQ, or from RQ to AQ file with the 7900HT Fast Real-Time PCR System?

SDS v2.4 and above allow you to convert your AQ files to RQ and vice versa.
To do so, simply go to: Tools then AQ RQ Converter
-Choose from the dropdown which direction you want to go (AQ to RQ, or RQ to AQ).
-Next click on ‘Add' and browse to the plate file location in the new window. and click ‘Open'.
-Click on ‘Convert'. A new file will be created in the same location as the original, but with _AQ or _RQ at the end of the filename.

If you do not have v2.4, you can upgrade your software here (http://www.thermofisher.com/us/en/home/technical-resources/software-downloads/applied-biosystems-7900ht-fast-real-timespcr-system.html).

One benefit of this conversion feature is that it allows you to convert 7900 files to the correct type to do analysis for relative quantification study, for example, if a plate is originally run as AQ instead of RQ, or if you would like to generate a standard curve quantification in the AQ if a plate was originally run as RQ.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Why are there no RQ values for all the samples in my study in the 7900HT Fast Real-Time PCR System software?

The 7900HT Fast Real-Time PCR System software requires that the endogenous control be run on every plate within a study. If not, you will only see RQ values for the plate that contains the endogenous control on it. The other plates will have Ct values for all the wells, but the software will not calculate RQ values. If you designed your experiments in this type of layout, you will want to do your analysis in the free DataAssist Software (https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/dataassist-software.html?icid=fr-realtimesoftware-5) instead, or use the Thermo Fisher Cloud (https://www.thermofisher.com/us/en/home/cloud.html). This program does not have the endogenous control requirement for every plate. Simply export the study results file from the 7900HT software as a *.txt or *.csv file, and use that to create a new study in the DataAssist software. You should now be able to see RQ values for all samples, even those without an endogenous control on the same plate.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Why do I not see any Ct or RQ values from my ddCT experiment in the 7900HT Fast Real-Time PCR System software?

In the 7900HT Fast Real-Time PCR System software, v2.3 or later, CT or RQ values are only provided in the RQ Manager software. RQ Manager is installed on the computer along with SDS v2.x.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What can I use for data analysis on the 7900HT Fast Real-Time PCR System?

For genotyping data, we recommend TaqMan Genotyper Software (https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/taqman-genotyper-software.html?icid=fr-software-1).
For relative quantitation, we recommend ExpressionSuite Software (https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/expressionsuite-software.html). You may also use the Thermo Fisher Cloud for either application. https://www.thermofisher.com/us/en/home/cloud.html

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What dyes can I use on the 7900HT Fast Real-Time PCR System?

The Applied Biosystems 7900HT Fast Real-Time PCR Systems use the following dye sets for calibration: FAM, SYBR, VIC, ROX, NED, TAMRA, JOE, TET dyes. Custom dyes can also be used that read between 500 nm and 660 nm, although you will have to calibrate the system first for any new dye.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.