(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals

CAS: 111769-27-8 | Molecular Formula: C11H17NO4S | Molecular Weight: 259.32 g/mol
Have Questions?
Catalog NumberQuantity
4358200505 g
Catalog number 435820050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS111769-27-8
IUPAC Name4-methylbenzene-1-sulfonic acid; oxolan-3-amine
Molecular FormulaC11H17NO4S
InChI KeyBZXPLADBSZWDIH-UHFFFAOYNA-N
SMILESNC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+3° to +5° (20°C, 589 nm) (c=1, methanol)
Elemental Analysis5.18 to 5.62 % N
Appearance (Color)White
Elemental Analysis48.91 to 52.99 % C
Appearance (Form)Crystalline powder
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only