Tower Computer Kit for 7300/7500/7500 Fast Real-Time PCR Systems - FAQs
Tower Computer Kit for 7300/7500/7500 Fast Real-Time PCR Systems - FAQs
View additional product information for Tower Computer Kit for 7300/7500/7500 Fast Real-Time PCR Systems - FAQs (4359284)
1 product FAQs found
Are there differences in the tube/plate format between Applied Biosystems 7500 Real-Time PCR Systems and Applied Biosystems 7500 Fast Real-Time PCR Systems?
Yes, the standard 7500 instrument has a 96 x 0.2 mL block, whereas the 7500 Fast instrument has a 96 x 0.1 mL block.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.