Notebook Computer Kit for 7300/7500/7500 Fast Real-Time PCR Systems - FAQs
Notebook Computer Kit for 7300/7500/7500 Fast Real-Time PCR Systems - FAQs
View additional product information for Notebook Computer Kit for 7300/7500/7500 Fast Real-Time PCR Systems - FAQs (4359286)
12 product FAQs found
Can TET dye be used on the Applied Biosystems real-time PCR instruments?
TET dye is not recommended to be used on the following real-time PCR instrument models: 7000, 7300, 7500, 7500 FAST, StepOne, and StepOnePlus Systems.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What dyes can I use as reporter dyes in a multiplex reaction on the Applied Biosystems 7300 Real-Time PCR Systems?
The Applied Biosystems 7300 Real-Time PCR System has 4 signal detection filters: Filter A (FAM and SYBR Green), Filter B (VIC and JOE), Filter C (TAMRA, NED and Cy3) and Filter D (ROX and Texas Red). For multiplexing reactions, the reporter dyes from the different assays should be chosen from different filter sets, so they can be compatible. In addition, the reporter dye need to be compatible with the quencher and the passive reference dye that might be present in the reaction. The commonly used combination is FAM and VIC.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What are the thermal cycling parameters of the Fast mode for the 7500 Fast Real-Time PCR System?
For the 7500 Fast real time PCR systems, the thermal cycling parameters are defaulted to 50°C/2 min for UNG incubation, 95°C/20 sec for fast polymerase activation, 40 cycles of 95°C/3 secs for the dissociation and 60°C/30 seconds for annealing and extensions.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I run slower, standard thermal cycling experiments on a 7500 Fast Real-Time PCR System?
Yes. you can choose either standard or fast ramp on the 7500 Fast System for software versions 2x or greater, and a 9600 emulation thermal cycling mode is available for customers who wish to run assays based on adjusted ramp rate. However, you should be aware that we cannot guarantee identical performance to the same modes run on a standard 7500 system block.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I compare the data collected when using Fast thermal cycling mode compared to using standard thermal cycling mode on the 7500 Fast Real-Time PCR System?
Runs can be compared between standard and fast mode, as the data collected are collected in all five filters in the instrument. However, keep in mind that these data may not always show identical results and may have added variability due to the data collection processes and differences in ramp rate between standard and fast mode.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
If I were to upgrade my existing 7500 System standard unit to the a 7500 Fast configuration, can I still open my old data files?
Yes, older 7500 or 7500 fast data files are compatible with the current version of the SDS software. However, there is one exception: please note that the SDS v1.3 software will not allow RQ Studies to be created from RQ Plates from different instrument types, different thermal modes (i.e., fast and standard modes) or different numbers of thermal cycles.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which computer operating systems are compatible with the7500 Fast Real-Time PCR System?
You can use up to Windows XP, Service Pack 3 (SP3) on the computer that is connected to the 7500 Fast Real-Time PCR System instrument. An exception is the SDS v2.0.5 software, which can be used with the Windows 7 operating system for stand-alone data analysis steps.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What is the recommended input template amount for each quantitative reaction on the 7500 Fast Real-Time PCR System?
The recommended input template range is 10-100 ng of gDNA/ cDNA template per reaction.
What is the recommended reaction volume that I can run for using 7500 Fast Real-Time PCR System?
We recommend running a total reaction volume between 10 and 30 ul per well.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
To analyze an RQ Study, can I import RQ plates from a combination of Fast and Standard mode runs on a 7500 Fast Real-Time PCR System?
We do not recommend this as the current and previous versions of SDS software will not allow RQ Studies to be created from RQ Plates from different instrument types, different thermal modes/profiles or different numbers of thermal cycles. It is important to reduce variability among RQ plates in an RQ study by importing RQ plates from the same instrument type, with the same thermal profile and mode, as well as the same block type and sample volume.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I run "standard" and "9600 emulation" modes on the 7500 Fast Real-Time PCR System?
Yes, standard and 9600 emulation modes are optionally available. We have matched the ramping rate and thus anticipate equivalent performance. However, the actual total run times will differ from standard and 9600 emulation modes run on a standard block, and we cannot guarantee identical results as performance may vary depending on the assay.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Are there differences in the tube/plate format between Applied Biosystems 7500 Real-Time PCR Systems and Applied Biosystems 7500 Fast Real-Time PCR Systems?
Yes, the standard 7500 instrument has a 96 x 0.2 mL block, whereas the 7500 Fast instrument has a 96 x 0.1 mL block.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.