m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
435971000100 mLAcroSeal™ Glass Bottle
Catalog number 435971000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS121905-60-0
IUPAC Namechloro(3-methylphenyl)magnesium
Molecular FormulaC7H7ClMg
InChI KeyBVYCQPCLCZTSPD-UHFFFAOYSA-M
SMILESCC1=CC([Mg]Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Yellow to red to brown
Titration Grignard0.95 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only