Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™
Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™
Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™
Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™
Thermo Scientific Chemicals

Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™

CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
435991000100 mLAcroSeal™ Glass Bottle
Catalog number 435991000
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Chemical Identifiers
CAS5574-97-0
IUPAC Nametetrabutylazanium dihydrogen phosphate
Molecular FormulaC16H38NO4P
InChI KeyARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILESOP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Colorless to light yellow
Titration with NaOH0.38 to 0.45 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only