n-Hexadecane, 99%, anhydrous, AcroSeal™
n-Hexadecane, 99%, anhydrous, AcroSeal™
n-Hexadecane, 99%, anhydrous, AcroSeal™
n-Hexadecane, 99%, anhydrous, AcroSeal™
n-Hexadecane, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

n-Hexadecane, 99%, anhydrous, AcroSeal™

CAS: 544-76-3 | C16H34 | 226.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 436141000
Price (TWD)
-
Request A Quote
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS544-76-3
IUPAC Namehexadecane
Molecular FormulaC16H34
InChI KeyDCAYPVUWAIABOU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid or solid
Infrared spectrumConforms
GC>=98.5 %
Color scale=<10 APHA
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only