Product Image
Applied Biosystems™

7500 Fast Real-Time PCR Systems Spectral Calibration Kit II

This easy-to-use kit establishes the pure dye spectra and multicomponent values for Cy3®, Cy5®, and Texas Red™ dyes on theRead more
Have Questions?
Catalog NumberQuantity
43622011 kit
Catalog number 4362201
Price (MXN)
-
Quantity:
1 kit
This easy-to-use kit establishes the pure dye spectra and multicomponent values for Cy3®, Cy5®, and Texas Red™ dyes on the 7500 Fast Real-Time PCR System. The Applied Biosystems® 7500 Real-Time PCR Systems Spectral Calibration Kit II contains three Optical 96-Well Fast Thermocycling Plates preloaded and sealed with Cy3®, Cy5®, and Texas Red™ fluorescent dye standards.

• 96-well optical reaction plates preloaded with Cy3®, Cy5®, and Texas Red™ dyes allow quick and easy calibration of the 7500 Fast Real-Time PCR System
• Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously

Spend Your Time on Research, Not Instrument Calibration
This kit's easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7500 Fast Real-Time PCR System. Insert the reaction plates during system installation, and the system collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeCy™3, Cy™5, Texas Red™
Calibration TypeSpectral Calibration
Detection MethodFluorescence
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7500 Fast System
Quantity1 kit
Shipping ConditionDry Ice
Product TypeSpectral Calibration Kit
Unit SizeEach
Contents & Storage
Contains three optical 96-Well Fast Thermocycling Plates preloaded and sealed, containing Cy3™ Texas Red™ and Cy5™ fluorescent dye standards.

Store at -15 to -25°C. Protect from light.

Expiration date: 12 months from the date of delivery