Dichloroisocyanuric acid sodium salt monohydrate, 97%
Dichloroisocyanuric acid sodium salt monohydrate, 97%
Dichloroisocyanuric acid sodium salt monohydrate, 97%
Thermo Scientific Chemicals

Dichloroisocyanuric acid sodium salt monohydrate, 97%

CAS: 52671-45-1 | C3Cl2N3NaO3·H2O | 237.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 436450500
Price (USD)
44.40
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
44.40
Each
Chemical Identifiers
CAS52671-45-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or granules
Titration Iodometric96.0 to 104.0 % (on dry substance)
Appearance (Color)White
Infrared spectrumConforms
Loss on drying6 % to 11 % (105°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only