Sulfamethoxypyridazine, Thermo Scientific Chemicals
Sulfamethoxypyridazine, Thermo Scientific Chemicals
Sulfamethoxypyridazine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfamethoxypyridazine, Thermo Scientific Chemicals

製品番号(カタログ番号)数量パッケージ
43647025025 gGlass bottle
製品番号(カタログ番号) 436470250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS80-35-3
IUPAC Name4-amino-N-(6-methoxypyridazin-3-yl)benzene-1-sulfonamide
Molecular FormulaC11H12N4O3S
InChI KeyVLYWMPOKSSWJAL-UHFFFAOYSA-N
SMILESCOC1=CC=C(NS(=O)(=O)C2=CC=C(N)C=C2)N=N1
さらに表示
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point180°C to 184°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only