(R)-(-)-3-Amino-1-BOC-piperidine, 97%, Thermo Scientific Chemicals
(R)-(-)-3-Amino-1-BOC-piperidine, 97%, Thermo Scientific Chemicals
(R)-(-)-3-Amino-1-BOC-piperidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-3-Amino-1-BOC-piperidine, 97%, Thermo Scientific Chemicals

CAS: 188111-79-7 | Molecular Formula: C10H20N2O2 | Molecular Weight: 200.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4364900505 gGlass bottle
Catalog number 436490050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS188111-79-7
IUPAC Nametert-butyl 3-aminopiperidine-1-carboxylate
Molecular FormulaC10H20N2O2
InChI KeyAKQXKEBCONUWCL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(N)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation-26.5° to -30.5° (20°C, 589 nm) (c=1, DMF)
Appearance (Color)Colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid or low melting solid
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only