TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG
Actual product may vary
Applied Biosystems™

TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG

TaqMan Fast Universal Master Mix (2X), No AmpErase UNG, delivers results in 40 minutes for 40 cycles of PCR inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
43660731250 Reactions
4366072500 Reactions
43678465000 Reactions
4352042250 Reactions
43641032500 Reactions
Catalog number 4366073
Price (USD)
1,774.65
Online exclusive
1,972.00
Save 197.35
Each
No. of Reactions:
1250 Reactions
Price (USD)
1,774.65
Online exclusive
1,972.00
Save 197.35
Each
TaqMan Fast Universal Master Mix (2X), No AmpErase UNG, delivers results in 40 minutes for 40 cycles of PCR in a 20-μL reaction volume using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems. The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup. Additionally, a proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments.

Convenience
Everything you need for TaqMan-based qPCR amplification and detection in a single-tube format. TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.

Compatibility
TaqMan Fast Universal PCR Master Mix (2X), No AmpErase UNG, is compatible with TaqMan Gene Expression assays.

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Fast Universal PCR Master Mix and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
FormatTube
No. of Reactions1250 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq DNA Polymerase
Product LineAmpErase, TaqMan
Product TypeFast Universal PCR Master Mix
Quantity1,250 reactions
Sample TypeDNA (Genomic), cDNA
Sufficient For250 Reactions
Volume1.25 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Genotyping, miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
This 5-pack contains five kits, each containing 2 tubes (1.25 ml each) of 2X TaqMan™ Fast Universal PCR Master Mix.

Store at 2°C to 8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

Is the TaqMan Fast Universal PCR Master Mix offered with AmpErase UNG?

The TaqMan Fast Universal PCR Master Mix is provided without any UNG and you should add in AmpEraseUNG to reactions separately, with the addition of an initial 2 minute cycling step at 50º C. If you prefer to have a master mix with the UNG pre-added, please use TaqMan Fast Advanced Master Mix.

Citations & References (43)

Citations & References
Abstract
Genetic analysis of hepatitis C virus with defective genome and its infectivity in vitro.
Authors:Sugiyama K, Suzuki K, Nakazawa T, Funami K, Hishiki T, Ogawa K, Saito S, Shimotohno KW, Suzuki T, Shimizu Y, Tobita R, Hijikata M, Takaku H, Shimotohno K
Journal:J Virol
PubMed ID:19369330
Replication and infectivity of hepatitis C virus (HCV) with a defective genome is ambiguous. We molecularly cloned 38 HCV isolates with defective genomes from 18 patient sera. The structural regions were widely deleted, with the 5' untranslated, core, and NS3-NS5B regions preserved. All of the deletions were in frame, ... More
Down Regulation of Genes Involved in T Cell Polarity and Motility during the Induction of Heart Allograft Tolerance by Allochimeric MHC I
Authors:Lisik, W; Tejpal, N; Gong, YQ; Skelton, TS; Ganachari, M; Bremer, EG; Kloc, M; Ghobrial, RM
Journal:
PubMed ID:
'Background: The allochimeric MHC class I molecule [α1h1/u]-RT1. Aa that contains donor-type (Wistar Furth, WF; RT1u) epitopes displayed on recipient-type (ACI, RT1a) administered in conjunction with sub-therapeutic dose of cyclosporine (CsA) induces indefinite survival of heterotopic cardiac allografts in rat model. In vascularized transplantation models, the spleen contributes to graft ... More
Activation of Macrophages by Polysaccharide-protein Complex from Lycium barbarum L.
Authors:Chen, ZS; Soo, MY; Srinivasan, N; Tan, BKH; Chan, SH
Journal:
PubMed ID:
'Macrophages play crucial roles in innate immunity. This paper reports that a polysaccharide-protein complex isolated from Lycium barbarum (LBP) is able to activate macrophages. LBP was isolated from Lycium barbarum fruit and separated to five homogenous fractions, designated LBPF1, LBPF2, LBPF3, LBPF4 and LBPF5. It was found that LBP (50 ... More
Fusion of Epstein-Barr virus nuclear antigen-1-derived glycine-alanine repeat to trans-dominant HIV-1 Gag increases inhibitory activities and survival of transduced cells in vivo
Authors:Hammer, D; Wild, J; Ludwig, C; Asbach, B; Notka, F; Wagner, R
Journal:
PubMed ID:
'Trans-dominant human immunodeficiency virus type 1 (HIV-1) Gag derivatives have been shown to efficiently inhibit late steps of HIV-1 replication in vitro by interfering with Gag precursor assembly, thus ranking among the interesting candidates for gene therapy approaches. However, efficient antiviral activities of corresponding transgenes are likely to be counteracted ... More
Prevalence of Chlamydophila psittaci in wild birds-potential risk for domestic poultry, pet birds, and public health?
Authors:Zweifel, D; Hoop, R; Sachse, K; Pospischil, A; Borel, N
Journal:
PubMed ID:
'To determine the prevalence of Chlamydophila psittaci in wild birds, cloacal swabs from 527 songbirds, 442 waterfowl, 84 feral pigeons, and 38 cormorants were examined by Chlamydiaceae-specific real-time polymerase chain reaction (PCR) and ArrayTube microarray assays for chlamydial species determination and genotyping of C. psittaci. Inconclusive cases were further characterized ... More