Applied Biosystems™

Plate Base for 3130 and 3100 Series Genetic Analyzers, 96 well, Fast (0.1 mL)

Catalog number:  4367470

 Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is an accessory part for use with the 3100- and 3130-series Genetic Analyzers.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 3100-Avant™ Genetic Analyzer, 3130 Genetic Analyzer
Format: 96-well plate
Product Size: 4 each
Shipping Condition: Room Temperature
Type: Base

Contents & storage

Each package contains 4 plate bases.

Documents

Manuals & protocols