PSI-CLONE™ 96-Well PCR Purification Plates

Catalog number: 4367833

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® PSI-Clone™ PCR 96-Well plates are for the rapid purification of PCR reaction products using either vacuum- or centrifugation-based protocols. Both protocols are fast and result in high-yield, high-purity DNA ideal for molecular biology experiments. 1 plate supplied.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

High Throughput Compatibility: High Throughput-Compatible
Sample Type (General): Enzymatic Reactions (e.g. PCR)
Downstream Application: Cloning
Format: 96-well plate
Product Line: PSI-CLONE™
Product Size: 1 plate
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols