Sodium carbonate, 99.999%, (trace metal basis)
Sodium carbonate, 99.999%, (trace metal basis)
Sodium carbonate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Sodium carbonate, 99.999%, (trace metal basis)

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 436800050
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.90
Save 10.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.90
Save 10.25
Each
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<10 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only