Pyrazinecarbonitrile, 99%
Pyrazinecarbonitrile, 99%
Pyrazinecarbonitrile, 99%
Thermo Scientific Chemicals

Pyrazinecarbonitrile, 99%

CAS: 19847-12-2 | C5H3N3 | 105.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 436920100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19847-12-2
IUPAC Namepyrazine-2-carbonitrile
Molecular FormulaC5H3N3
InChI KeyPMSVVUSIPKHUMT-UHFFFAOYSA-N
SMILESN#CC1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow
Appearance (Form)Solid or liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only