Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

数量:
25 g
100 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 436961000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6484-52-2
IUPAC Nameammonium nitrate
Molecular FormulaH4N2O3
InChI KeyDVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES[NH4+].[O-][N+]([O-])=O
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline powder
Total trace metal impurities=<10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only