3130 Series Training DVD

Catalog number: 4370107

Applied Biosystems™  Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 3130 Series Training DVD covers instrument maintenance and software set up for Sequencing Analysis Software, GeneMapper® Software and Seqscape® Software.

The 3130 Genetic Analyzer is the latest generation of 4-capillary electrophoresis instruments for the low to medium throughput laboratories. The system offers industry-leading performance, plus sophisticated automation capabilities allowing you to save time, reduce costs and increase productivity.

With the Applied Biosystems® 3130 Genetic Analyzer, you get the sophisticated automation and superior performance of the 3130 system, at acquisition and operating costs tailored to a growing research lab.
• Run a wide variety of sequencing and fragment analysis applications including microsatellite analysis, AFLP, LOH, SNP validation, and SNP screening.
• Reduce maintenance time by eliminating manual syringe washing and filling with automated polymer delivery.
• Increase your data quality for sequencing and fragment analysis applications – longer read length, and higher resolution with shorter run times.
• Increase laboratory productivity and turn around time by processing 96- or 384-well plates with a four-capillary array.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition: Room Temperature
Product Line: Applied Biosystems™
Product Size: 1 kit

Contents & storage

Contains one DVD.

Documents

Manuals & protocols