Thermo Scientific Chemicals

Potassium tetrachloroplatinate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437030010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium tetrachloroplatinate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 437030010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 437030010
Price (USD)
Price: 161.00
Online price: 136.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10025-99-7
IUPAC Name
dipotassium tetrachloroplatinumdiuide
Molecular Formula
Cl4H4K2Pt
InChI Key
JCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILES
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
Appearance (Color)
Orange to red to brown
Total trace metal impurities
=<100 ppm
Appearance (Form)
Crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text