SurePac Oligo RP MDI is a robust reversed-phase column designed for oligonucleotide analysis by HPLC and LC-MS. It delivers high recovery, bioinert column hardware, and reproducible separations, enabling reliable profiling of oligonucleotides of any size.
Have Questions?
變更視圖
產品號碼
Diameter (Metric)
Length (Metric)
43712-102132
2.1 mm
100 mm
43712-022132
2.1 mm
20 mm
43712-052132
2.1 mm
50 mm
43712-050332
300 μm
50 mm
43712-0510032
10 mm
50 mm
43712-0521232
21.2 mm
50 mm
6 選項
產品號碼 43712-102132
價格 (TWD)
-
Diameter (Metric):
2.1 mm
Length (Metric):
100 mm
Thermo Scientific™ SurePac™ Oligo RP MDi™ HPLC columns deliver high-resolution, reproducible separations for therapeutic oligonucleotides and nucleic acids across R and D and QC workflows. Built on monodisperse, supermacroporous polymeric packing with bio-inert hardware, they provide stable performance even under elevated pH and temperature conditions, resolving truncation impurities while supporting large oligos and intact nucleic acids for scalable LC-MS and LC-UV analysis.
Purpose-built for oligonucleotide RP separations
Optimized for DNA and RNA oligonucleotides, including modified sequences, delivering consistent retention and selectivity across short and long oligos.
Bioinert, metal-free fluid path
Reduces metal/oligonucleotide interactions to improve recovery, minimize adsorption, and deliver cleaner peak shapes for sensitive analyses.
Optimized for ion-pair RP LC-MS
Fully compatible with common ion-pairing reagents such as HFIP/TEA, enabling sensitive, reproducible LC-MS detection.
High recovery of long and modified oligonucleotides
High resolution power for oligonucleotides spanning over a wide size range from small to (ultra) large
Supports efficient analysis of long, hydrophobic, and chemically complex oligos without compromising signal or resolution.
Robust stationary phase for demanding methods
Maintains stable performance under high organic content and elevated temperatures, supporting fast gradients and long column lifetimes.
Excellent peak shape and impurity resolution
Delivers sharp, symmetrical peaks and reliable separation of closely related species, including impurities and sequence variants.