Product Image
Applied Biosystems™
MicroSEQ™ ID Microbial Identification Software v2.0 Bundle (MSID v2.0 Analysis Software with Bacterial & Fungal Libraries)
MicroSEQ™ ID v2.1.1 is an analysis application used to basecall, assemble, trim, and analyze the data generated using the MicroSEQ™Read more
Have Questions?
Catalog NumberQuantity
43712981 Kit
Catalog number 4371298
Price (USD)
53,000.00
Each
Quantity:
1 Kit
Price (USD)
53,000.00
Each
MicroSEQ™ ID v2.1.1 is an analysis application used to basecall, assemble, trim, and analyze the data generated using the MicroSEQ™ Microbial Identification Kits. The software allows the user to create consensus sequences from acquired data, and compare the consensus to an existing library of known sequences. This software package includes the MicroSEQ™ ID 16S rDNA 500, 16S rDNA Full Gene, and the D2 Fungal libraries.
MicroSEQ™ ID Analysis Software enables you to identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library.
• A validated library of over 2300 microbial sequences ensures you get the answers you need.
• Easy-to-use interface and optimized search algorithms enable quick results.
• Phylogenetic trees provide a deeper understanding of your unknowns.
• Security features support 21 CFR Part 11 compliance, enabling the software to integrate smoothly into your workflow.
Confidently Identify Microbes
The MicroSEQ™ system comprises bacterial libraries of 500 bp sequences and of over 1,400 validated entries--including extensive coverage of gram-negative nonfermenters, Bacillus, Coryneforms, Mycobacteria, and Staphylococcus. Another library containing more than 900 entries is available for fungal identification. To ensure you can accurately identify the species of your sequences, we rigorously validate all library entries. And unlike other widely available databases, MicroSEQ™ libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections. The libraries also support sequence polymorphisms, and include bacterial Category A biothreat agents.
Flexible and Easy-to-Use
To simplify microbial sequence identification, analysis, and reporting, MicroSEQ™ ID Analysis Software provides easy-to-use wizards and a powerful, automated toolset. The software also includes a full complement of audit-trail and security features that support 21 CFR Part 11 compliance. Further, this flexible software even allows you to create customized entries for specific sequences from new or proprietary strains.
The MicroSEQ™ System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems MicroSEQ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
Product LineMicroSEQ
Product TypeID Microbial Identification Software V3.0 Bundle
Quantity1 Kit
Unit SizeEach