Silicon dioxide, 99.998%, (trace metal basis)
Silicon dioxide, 99.998%, (trace metal basis)
Silicon dioxide, 99.998%, (trace metal basis)
Thermo Scientific Chemicals

Silicon dioxide, 99.998%, (trace metal basis)

For ultra trace analysis | CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
20 g
100 g
Packaging:
Glass bottle
Catalog number 437150050
Price (EUR)
40,65
Online exclusive
44,90
Save 4,25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
40,65
Online exclusive
44,90
Save 4,25
Each
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<20 ppm
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Assay44.5 to 47.9 % (Si)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only