3-Cyano-4-iodopyridine, 97%
3-Cyano-4-iodopyridine, 97%
3-Cyano-4-iodopyridine, 97%
Thermo Scientific Chemicals

3-Cyano-4-iodopyridine, 97%

CAS: 490039-72-0 | C6H3IN2 | 230.01 g/mol
製品番号(カタログ番号) 437450050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS490039-72-0
IUPAC Name4-iodopyridine-3-carbonitrile
Molecular FormulaC6H3IN2
InChI KeyCJKHIIQONPNAIP-UHFFFAOYSA-N
SMILESIC1=C(C=NC=C1)C#N
さらに表示
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder
Melting point112°C to 116°C
Appearance (Color)Light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only