Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates
Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates
Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates
Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates
Thermo Scientific™

Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates

Use these well plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays.
Have Questions?
Catalog NumberColor
437796White
Catalog number 437796
Price (USD)
364.59
Online exclusive
478.07
Save 113.48
Case of 60
Color:
White
Price (USD)
364.59
Online exclusive
478.07
Save 113.48
Case of 60

Use Thermo Scientific™ Nunc FluoroNunc/LumiNunc 96-Well Solid plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays. Thermo Scientific Nunc Immuno plates offer lot-to-lot reliability, well-to-well reproducibility, and are backed by industry-leading quality standards and certifications.

  • Plate has SBS footprint—fits standard equipment
  • Available with flat-bottom or C-shaped wells
  • Choice of PolySorp™ or MaxiSorp™ surface
  • Offered as black, white or clear plates
  • Certified binding homogeneity/reproducibility

Choice of Surface

  • PolySorp - Hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains

  • MaxiSorp - Hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character. Available with or without lot specific

  • performance testing via an IgG binding assay (Well to Well %CV less than 5% for clear plates and less than 10% for colored plates). Tested items come with a certificate

White

  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence

Black

  • Minimizes background in fluorescence reading
  • Minimizes back-scattered light
  • Minimizes crosstalk

Clear

  • For time-resolved fluorescence

Compatible with:

Compatible with lids for MicroWell™ plates, sealing tape and breathable membrane

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Binding PropertyHydrophilic
Binding TypeAdsorption
Certifications/ComplianceCertified for the absorption of IgG; well to well %CV of less than 10%.
Culture Area2.5 cm2
Detection MethodChemiluminescence
FormatStandard
LidWith Lid
No. per Case60
No. per Pack5
Packaging60 Cases
Plate BlockingNone
Product TypeMicroplate
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Working250 μL
Array8 x 12
ColorWhite
MaterialPolystyrene
No. of Wells96
Product LineNunc
Surface TreatmentMaxiSorp
Well ShapeRound, C Bottom
Unit SizeCase of 60

Frequently asked questions (FAQs)

What length of peptide is ideal for binding to the Thermo Scientific MaxiSorp surface? What are the detection limitations?

We have tested and found that a 3 amino acid peptide (Pro, Leu, Gly) cannot be detected when passively adsorbed on the MaxiSorp surface. However, this peptide can be detected when covalently immobilized using CovaLink NH Modules and CovaLink NH2 Modules and Plates. Using covalent immobilization of small peptide residues, you can typically obtain a better orientation of the molecule and reduced problems with antibody recognition of the peptide due to masking of the epitope. We have discovered that a 7 amino acid peptide from the MHC Class II antigen can be detected when adsorbed on the MaxiSorp surface. We state that the detection limitation using the MaxiSorp surface is between 3 and 7 amino acid residues.
One additional note is that detection is contingent upon the orientation of the peptide when immobilized. If the active site is inactivated or hidden at the site facing the solid phase, no detection signal is observed.

Can I bind either single- or double-stranded DNA to the Thermo Scientific MaxiSorp surface?

Single-stranded DNA can be adsorbed to our MaxiSorp surface using approximately 10 µg ssDNA per mL PBS, pH 8.2, although the stability is uncertain. Based on our experience, ssDNA immobilized on the MaxiSorp surface is so loosely bound that it can be removed by stringent washing.
Double-stranded DNA will not bind to the MaxiSorp surface. DNA, however, can be covalently bound to Nunc NucleoLink Strips.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

What is the difference between certified and non-certified MaxiSorp plates and modules?

Both of these surfaces are identical. The only difference between them is that for the certified plates, a representative sample from each manufacturing lot undergoes a Binding Capacity test. This test is an ELISA-like assay used in our quality control laboratories to ensure binding capabilities.

How can I reduce high background readings and non-specific binding, when performing ELISA using Nunc-Immuno MaxiSorp Plates?

Assay sensitivity depends strongly on efficient removal of non-specific reacting molecules. High background readings and coating instability can be minimized by addition of a blocking step after the first coating. The excess surface is then occupied by indifferent molecules.
We recommend washing three times after each coating step by using a solution of 0.15 M phosphate buffer (pH 7.2) with 0.2 M NaCl and 0.05% Tween 20.
For blocking, we recommend using 0.5% BSA, 1% casein or 1% gelatin in 0.15 M phosphate buffer (pH 8.2) or carbonate buffer (pH 9.6).