Cyclopentanesulfonyl chloride, 90%
Cyclopentanesulfonyl chloride, 90%
Cyclopentanesulfonyl chloride, 90%
Thermo Scientific Chemicals

Cyclopentanesulfonyl chloride, 90%

CAS: 26394-17-2 | C5H9ClO2S | 168.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 437830010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26394-17-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=88.0 %
Appearance (Form)Liquid
Appearance (Color)Light brown to dark brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only