The Applied Biosystems™ TaqMan™ Rat Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis ofRead more
Have Questions?
Catalog Number
Quantity
4378706
4 cards
Catalog number 4378706
Price (USD)
2,635.00
Each
Quantity:
4 cards
Price (USD)
2,635.00
Each
The Applied Biosystems™ TaqMan™ Rat Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 48 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.
The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
Research CategoryCell Cycle & Proliferation, Oncology, Toxicology & Drug Metabolism, Neurobiology, Inflammation
SpeciesRat
TargetIncludes five distinct phosphodiesterase families
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored (at 4°C.)
Frequently asked questions (FAQs)
What are the storage recommendations for TaqMan Array Cards (TAC)?
We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.