Product Image
Invitrogen™

Tempus™ Spin RNA Isolation Kit

This Applied Biosystems™ product is for the fast, easy, glass fiber filter–based isolation of RNA from blood using standard laboratoryRead more
Have Questions?
Catalog NumberQuantity
438020450 Preps
Catalog number 4380204
Price (ZAR)
-
Quantity:
50 Preps
This Applied Biosystems™ product is for the fast, easy, glass fiber filter–based isolation of RNA from blood using standard laboratory equipment in about one hour. Contains sufficient reagents for RNA purification from 50 samples.

• Isolate high-quality RNA from blood, suitable for real-time PCR and microarray analysis
• Obtain highly stable RNA virtually free of RNase activity, contaminating gDNA, or RT-PCR inhibitors
• Easy workflow requires only standard laboratory centrifuges and ∼1 hour of time
• No separate RBC lysis or proteinase K treatment required

Gene expression profiling and analysis of whole blood is becoming an increasingly important life science tool. Accurately measuring the molecular profile of blood-based RNA transcripts is a critical need in areas of research such as disease association studies, biomarker discovery and validation, and monitoring of drug treatment regimes.

Accessory Product
The highly unstable nature of RNA transcripts ex vivo creates an even greater challenge for accurate expression analysis. Ongoing metabolic changes can drastically affect gene expression profiles in as little as one hour in standard blood collection tubes (see Figure). Use of the Applied Biosystems™ Tempus™ Blood RNA Tubes (SKU #4342792) is therefore recommended. These tubes are designed for direct collection of 3 mL of blood into a plastic, evacuated blood collection tube containing RNA Stabilizing Reagent. The RNA, and therefore the expression profile, is stable in the collection tube for up to five days at room temperature and minimally 7 days at 4°C (see Figure).

Performance with Ease
The ability of the Tempus™ system (Tempus™ Blood RNA Tubes and Tempus™ Spin RNA Isolation Kit) to stabilize RNA expression profiles has been tested over a range of storage conditions (see Figure). The combination provides stable RNA, high yields of purified RNA (observed yields of 6–25 μg from a full blood draw from normal, healthy donors), and high-quality RNA (A260/280 ratio greater than 1.9 and RIN greater than 7.0) suitable for use in downstream applications such as quantitative PCR, microarray profiling, and other RNA analysis methods.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume100 μL
Final Product TypeTotal RNA
For Use With (Application)RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Purification Time40 min.
Quantity50 Preps
Shipping ConditionRoom Temperature
Starting Material AmountUp to 3 mL of blood
Yield≤25 μg
Isolation TechnologySilica Spin Column
Sample TypeWhole Blood
Unit SizeEach
Contents & Storage
• 50 RNA Purification Filters; room temperature
• 2 x 100 Collection Tubes; room temperature
• 2 x 80 mL 1X PBS; room temperature
• 24 mL RNA Purification Resuspension Solution; room temperature
• 2 x 22 mL RNA Purification Wash Solution 1; room temperature
• 120 mL RNA Purification Wash Solution 2; room temperature
• 4 x 1.9 mL Nucleic Acid Purification Elution Solution; room temperature

Frequently asked questions (FAQs)

Will the Tempus system work to isolate miRNA?

No, unfortunately, small RNAs will be lost using this purification method due to the low molecular weight cutoff from the column (approximately 200 bp).

I am doing blood work and using the Tempus collection tubes. Is the Tempus system compatible with the RiboPure-Blood RNA Purification Kit?

No, please follow the RNA isolation methods described with the Tempus system. The collection tubes are not compatible with the RiboPure kit.

What is the recommended storage temperature and stability of samples in the Tempus Blood RNA Tube?

Our general recommendations are that once blood has been collected in the Tempus Blood RNA Tubes, you can store the tube at room temperature for up to 5 days or at 4 degrees C for up to 7 days. For long term storage, the tube can be frozen at -80 degrees C for at least 1.5 years.

Can I add a DNase step to the RNA isolation protocol in the Tempus Spin RNA Isolation Kit?

Yes, it is okay to add an on-column or post-elution DNase treatment.

For the on-column DNase treatment, use 350 µl of the following solution and incubate for 15 mins at room temperature:

2500 units DNAse I (RNAse-free) in 10 mL of solution containing:
10 mM Tris-HCl, pH 8
2.5 mM MgCl2 . 6H2O
0.1 mM CaCl2 . 2H2O

This would be done between the two Wash 2 steps, and it would be a good idea to add a third Wash 2 step to make sure all of the DNase is washed away.