NativePac OBE-1 SEC Column for Online Sample Preparation
Thermo Scientific™

NativePac OBE-1 SEC Column for Online Sample Preparation

Perform Native-MS analysis of biomacromolecules with the NativePac OBE-1 SEC Column designed for rapid exchange of non-volatile buffer components with a volatile buffer.

Have Questions?
Catalog NumberLength (Metric)
43803-05213050 mm
Catalog number 43803-052130
Price (USD)
358.82
Each
Length (Metric):
50 mm
Price (USD)
358.82
Each

The Thermo Scientific™ NativePac OBE-1 Column is a Size Exclusion Chromatography (SEC) column designed for rapid exchange of non-volatile buffer components with a volatile buffer for Native-MS analysis of biomacromolecules. The small pores of the SEC stationary phase maximize separation of the non-volatile components from the sample of interest while the short column length minimizes analysis times to less than 2 minutes. A narrow column ID enables the use of low flow rates for high sensitivity detection and maximizes up time by minimizing buffer consumption.

The NativePac OBE-1 SEC column for online sample preparation offers the following features:

  • OBE column for protein molecular weight and structure screening
  • Rapid exchange of non-volatile buffer components with a volatile buffer for Native-MS analysis
  • Short column length minimizes analysis times and buffer consumption
  • Narrow column ID enables low flow rates for high sensitivity
  • Small SEC stationary phase pore size maximizes separation of non-volatile components from samples of interest
Specifications
Column FormatAnalytical Column
Column TypeOnline Buffer Exchange
Max. Pressure1450 psi (100 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Particle ShapeSpherical
Stationary PhaseProprietary Diol
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Particle Size3 μm
PhaseSize Exclusion
Pore Size80 Å
pH2 to 8
Unit SizeEach